Anti-Glucosamine Rabbit Polyclonal Antibody
Lieferant: US Biological
G3040-89A
USBIG3040-89AEA 649 EUR
USBIG3040-89A
Anti-Glucosamine Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: Glucosamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human