Anti-Glucose Transporter type 1 Rabbit Polyclonal Antibody
Lieferant: US Biological
G3900-03E
USBIG3900-03EEA 822 EUR
USBIG3900-03E
Anti-Glucose Transporter type 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:500; Western Blot: 1:1000
Type: Primary
Antigen: Glucose Transporter type 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human