Anti-GPX4 Mouse Monoclonal Antibody [clone: 10K213]

Lieferant: US Biological
G8127-14D
USBIG8127-14DEA 479 EUR
USBIG8127-14D
Anti-GPX4 Mouse Monoclonal Antibody [clone: 10K213]
Antikörper
Recommended dilution: Immunoprecipitation: 1-2?l/400?l s

Type: Primary
Antigen: GPX4 (glutathione peroxidase 4)
Clonality: monoclonal
Clone: 10K213
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR