Anti-Glyceraldehyde 3 Phosphate Dehydrogenase Goat polyclonal antibody

Lieferant: US Biological
G8140-01D
USBIG8140-01DEA 551 EUR
USBIG8140-01D
Anti-Glyceraldehyde 3 Phosphate Dehydrogenase Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:2000; Western Blot: 0. 01-0. 03?g/ml; Immunochemistry: 0. 3?g/ml

Type: Primary
Antigen: Glyceraldehyde 3 Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR