Anti-Glyceraldehyde 3 Phosphate Dehydrogenase Mouse monoclonal antibody [clone: 9L275]

Lieferant: US Biological
G8140-01H
USBIG8140-01HEA 767 EUR
USBIG8140-01H
Anti-Glyceraldehyde 3 Phosphate Dehydrogenase Mouse monoclonal antibody [clone: 9L275]
Antikörper
Type: Primary
Antigen: Glyceraldehyde 3 Phosphate Dehydrogenase
Clonality: monoclonal
Clone: 9L275
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Dog, Goat, Human, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR