Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Goat polyclonal antibody

Lieferant: US Biological
G8140-01P
USBIG8140-01PEA 697 EUR
USBIG8140-01P
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Goat polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 0. 3?g/ml; ELISA: 1:2000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR