Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Sheep polyclonal antibody

Lieferant: US Biological
G8140-02D
USBIG8140-02DEA 513 EUR
USBIG8140-02D
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Sheep polyclonal antibody
Antikörper
Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: E. coli, Human, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR