Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody
Lieferant: US Biological
G8140-03B
USBIG8140-03BEA 424 EUR
USBIG8140-03B
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat