Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Mouse Monoclonal Antibody [clone: 7C33]

Lieferant: US Biological
G8140-14B-100
USBIG8140-14B-100EA 776 EUR
USBIG8140-14B-100
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Mouse Monoclonal Antibody [clone: 7C33]
Antikörper
Recommended dilution: Western Blot: 3-6?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: Monoclonal
Clone: 7C33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken, Frog, Human, Mouse, Rabbit,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR