Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody

Lieferant: US Biological
G8140-16N
USBIG8140-16NEA 673 EUR
USBIG8140-16N
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Avian, Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR