Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Mouse monoclonal antibody [clone: 8L49]

Lieferant: US Biological
G8140-16P
USBIG8140-16PEA 485 EUR
USBIG8140-16P
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase Mouse monoclonal antibody [clone: 8L49]
Antikörper
Recommended dilution: Western Blot: 1:20000

Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase
Clonality: monoclonal
Clone: 8L49
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR