Anti-Glycine Receptor alpha 1/2 Rabbit polyclonal antibody

Lieferant: US Biological
G8167-02
USBIG8167-02EA 802 EUR
USBIG8167-02
Anti-Glycine Receptor alpha 1/2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 1:1000; Western Blot: 1:1000

Type: Primary
Antigen: Glycine Receptor alpha 1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR