Anti-Glycogen Phosphorylase Isoenzyme BB Mouse monoclonal antibody [clone: 6F1]

Lieferant: US Biological
G8170-02A
USBIG8170-02AEA 559 EUR
USBIG8170-02A
Anti-Glycogen Phosphorylase Isoenzyme BB Mouse monoclonal antibody [clone: 6F1]
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Glycogen Phosphorylase Isoenzyme BB
Clonality: monoclonal
Clone: 6F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR