Anti-Glyoxal Rabbit Polyclonal Antibody (Unconjugated)

Lieferant: US Biological
G8235-29
USBIG8235-29EA 795 EUR
USBIG8235-29
Anti-Glyoxal Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Glyoxal
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR