Anti-G Protein Coupled Receptor 20 Rabbit polyclonal antibody

Lieferant: US Biological
G8600-24B
USBIG8600-24BEA 681 EUR
USBIG8600-24B
Anti-G Protein Coupled Receptor 20 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: G Protein Coupled Receptor 20
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR