Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 10C424]

Lieferant: US Biological
G8900-01C
USBIG8900-01CEA 598 EUR
USBIG8900-01C
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 10C424]
Antikörper
Type: Primary
Antigen: Gram Negative Endotoxin
Clonality: monoclonal
Clone: 10C424
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR