Anti-Guanine Nucleotide Binding Protein, beta 4 subunit Chicken Polyclonal Antibody

Lieferant: US Biological
G9039-31
USBIG9039-31EA 608 EUR
USBIG9039-31
Anti-Guanine Nucleotide Binding Protein, beta 4 subunit Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: Guanine Nucleotide Binding Protein, beta 4 subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR