Anti-Guanine Nucleotide Binding Protein, beta 5 subunit Rabbit Polyclonal Antibody
Lieferant: US Biological
G9039-35
USBIG9039-35EA 701 EUR
USBIG9039-35
Anti-Guanine Nucleotide Binding Protein, beta 5 subunit Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: Guanine Nucleotide Binding Protein, beta 5 subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse