Anti-guanine nucleotide binding protein, beta 5 Subunit Rabbit Polyclonal Antibody

Lieferant: US Biological
G9039-35A
USBIG9039-35AEA 695 EUR
USBIG9039-35A
Anti-guanine nucleotide binding protein, beta 5 Subunit Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: guanine nucleotide binding protein, beta 5 Subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR