Anti-Guanine Nucleotide Binding Protein G Rabbit Polyclonal Antibody

Lieferant: US Biological
G9039-42
USBIG9039-42EA 633 EUR
USBIG9039-42
Anti-Guanine Nucleotide Binding Protein G Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-1:1000; Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Guanine Nucleotide Binding Protein G
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR