Anti-HCK Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: US Biological
H1827-05-FITC
USBIH1827-05-FITCEA 884 EUR
USBIH1827-05-FITC
Anti-HCK Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: HCK (hemopoietic cell kinase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human