Anti-HSPD1 Mouse Monoclonal Antibody [clone: 4E100]

Lieferant: US Biological
H1830-77A-200
USBIH1830-77A-200EA 677 EUR
USBIH1830-77A-200
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 4E100]
Antikörper
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: monoclonal
Clone: 4E+100
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Dog, Drosophila, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR