Anti-Heparan Sulfate Mouse Monoclonal Antibody [clone: 8.S.087]

Lieferant: US Biological
H1890
USBIH1890EA 557 EUR
USBIH1890
Anti-Heparan Sulfate Mouse Monoclonal Antibody [clone: 8.S.087]
Antikörper
Recommended dilution: Flow Cytometry: 1:70-1:140; Immunochemistry: 1:35-1:70

Type: Primary
Antigen: Heparan Sulfate
Clonality: monoclonal
Clone: 8.S.087
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR