Anti-Hepatitis B Surface Antigen PreS2 Rabbit Polyclonal Antibody

Lieferant: US Biological
H1915-98G
USBIH1915-98GEA 682 EUR
USBIH1915-98G
Anti-Hepatitis B Surface Antigen PreS2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Hepatitis B Surface Antigen PreS2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR