Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: 10D99]

Lieferant: US Biological
H1920-12C
USBIH1920-12CEA 652 EUR
USBIH1920-12C
Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: 10D99]
Antikörper
Recommended dilution: ELISA: 1:20-1:200; Western Blot: 1:10-1:50; Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Hepatitis C Virus Core Antigen
Clonality: monoclonal
Clone: 10D99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR