Anti-HIF1AN Rabbit Polyclonal Antibody
Lieferant: US Biological
H3848-85
USBIH3848-85EA 614 EUR
USBIH3848-85
Anti-HIF1AN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human