Anti-HIF1AN Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
H3848-85B-AP
USBIH3848-85B-APEA 884 EUR
USBIH3848-85B-AP
Anti-HIF1AN Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:250-1:500
Do not freeze!

Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR