Anti-HIF1AN Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
H3848-85B-PE
USBIH3848-85B-PEEA 884 EUR
USBIH3848-85B-PE
Anti-HIF1AN Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250-1:500
Do not freeze!

Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR