Anti-HIF1AN Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
H3848-85C-BIOT
USBIH3848-85C-BIOTEA 884 EUR
USBIH3848-85C-BIOT
Anti-HIF1AN Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR