Anti-HIF1AN Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
H3848-85C-FITC
USBIH3848-85C-FITCEA 884 EUR
USBIH3848-85C-FITC
Anti-HIF1AN Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250-1:500
Do not freeze!

Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR