Anti-HIF1AN Rabbit Monoclonal Antibody [clone: 10E26]
Lieferant: US Biological
H3848-85H
USBIH3848-85HEA 608 EUR
USBIH3848-85H
Anti-HIF1AN Rabbit Monoclonal Antibody [clone: 10E26]
Antikörper
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: monoclonal
Clone: 1E+27
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat