Anti-Histamine Receptor 3 Rabbit polyclonal antibody
Lieferant: US Biological
H5090-20-100
USBIH5090-20-100EA 742 EUR
USBIH5090-20-100
Anti-Histamine Receptor 3 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: Histamine Receptor 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human