Anti-HDAC2 Rabbit polyclonal antibody
Lieferant: US Biological
H5109-47N
USBIH5109-47NEA 559 EUR
USBIH5109-47N
Anti-HDAC2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: HDAC2 (histone deacetylase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human