Anti-H2AFX Rabbit Polyclonal Antibody

Lieferant: US Biological
H5110-03Y
USBIH5110-03YEA 449 EUR
USBIH5110-03Y
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:2500; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR