Anti-H2AFX Rabbit Polyclonal Antibody

Lieferant: US Biological
H5110-08A1
USBIH5110-08A1EA 964 EUR
USBIH5110-08A1
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunoblot: A 1:2000-1:10000

Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR