Anti-Histone H3, Citrulline 2, 8, 17 Rabbit polyclonal antibody

Lieferant: US Biological
H5110-12Q
USBIH5110-12QEA 711 EUR
USBIH5110-12Q
Anti-Histone H3, Citrulline 2, 8, 17 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:200-1:1000

Type: Primary
Antigen: Histone H3, Citrulline 2, 8, 17
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR