Anti-Histone H3, di-trimethyl K27 Mouse monoclonal antibody [clone: 9F617]

Lieferant: US Biological
H5110-14S6
USBIH5110-14S6EA 786 EUR
USBIH5110-14S6
Anti-Histone H3, di-trimethyl K27 Mouse monoclonal antibody [clone: 9F617]
Antikörper
Recommended dilution: Immunofluorescence: 1:1000; ELISA: 1:1000

Type: Primary
Antigen: Histone H3, di-trimethyl K27
Clonality: monoclonal
Clone: 9F617
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR