Anti-HIV-1 Goat Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
H6001-10C
USBIH6001-10CEA 478 EUR
USBIH6001-10C
Anti-HIV-1 Goat Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: HIV-1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR