Anti-HLA Class 2 Antigen DP, DQ, DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10D177]

Lieferant: US Biological
H6099-85E1
USBIH6099-85E1EA 607 EUR
USBIH6099-85E1
Anti-HLA Class 2 Antigen DP, DQ, DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10D177]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DP, DQ, DR
Clonality: monoclonal
Clone: 10D177
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR