Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: MEM-136]
Lieferant: US Biological
H6099-85L
USBIH6099-85LEA 607 EUR
USBIH6099-85L
Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: MEM-136]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DP, DR
Clonality: monoclonal
Clone: MEM-136
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human