Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: HL-40]

Lieferant: US Biological
H6099-85M
USBIH6099-85MEA 607 EUR
USBIH6099-85M
Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: HL-40]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DP, DR
Clonality: monoclonal
Clone: HL-40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR