Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3H2224]

Lieferant: US Biological
H6099-95G
USBIH6099-95GEA 561 EUR
USBIH6099-95G
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3H2224]
Antikörper
Do not freeze!

Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 3H2224
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR