Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F62]

Lieferant: US Biological
H6099-95K
USBIH6099-95KEA 350 EUR
USBIH6099-95K
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F62]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 5F62
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR