Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10D419]

Lieferant: US Biological
H6099-95Z1
USBIH6099-95Z1EA 916 EUR
USBIH6099-95Z1
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10D419]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 10D419
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR