Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 10D183]
Lieferant: US Biological
H6099-96H
USBIH6099-96HEA 713 EUR
USBIH6099-96H
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 10D183]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 10D183
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human