Anti-HLA Class 2 Antigen DR beta Mouse Monoclonal Antibody [clone: 8L293]

Lieferant: US Biological
H6099-96N
USBIH6099-96NEA 711 EUR
USBIH6099-96N
Anti-HLA Class 2 Antigen DR beta Mouse Monoclonal Antibody [clone: 8L293]
Antikörper
Recommended dilution: Flow Cytometry: 1:10-1:1000; ELISA: 1:100-1:2000; Western Blot: 1:100-1:2000; Immunochemistry: 1:10-1:500

Type: Primary
Antigen: HLA Class 2 Antigen DR beta
Clonality: monoclonal
Clone: 8L293
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR