Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: HL39]

Lieferant: US Biological
H6099-96R
USBIH6099-96REA 628 EUR
USBIH6099-96R
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: HL39]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: HL39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR