Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: L243]

Lieferant: US Biological
H6100-42L
USBIH6100-42LEA 338 EUR
USBIH6100-42L
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: L243]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: L243
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR