Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: LN3]

Lieferant: US Biological
H6100-42Q
USBIH6100-42QEA 357 EUR
USBIH6100-42Q
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: LN3]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: LN3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR