Anti-Homocysteine Rabbit Polyclonal Antibody

Lieferant: US Biological
H6950-04-100
USBIH6950-04-100EA 799 EUR
USBIH6950-04-100
Anti-Homocysteine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Homocysteine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR